Rescue Critters® Goldie Breath/Heart Sounds Simulator
Supplier: Rescue Critters
350
470126-440EA 5140.09 CAD
470126-440
Rescue Critters® Goldie Breath/Heart Sounds Simulator
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Made For Veterinary Auscultation
  • Changeable breath sounds for student review
  • Multiple speakers allow for specific placement
  • Made to order
Instructors can chose various sound scenarios for the classroom. This product uses plug-in modules, with data collated from actual patients. Five speakers are installed directly in our Goldie canine BHS manikin. The BHS unit also has two lights that illuminate during expiration. It is powered by a 9V battery and comes with a limited warranty.
Each Goldie comes with a tutor MS, one Breath Sound module, one Heart Sound module, five installed custom speakers and a 9V battery.
Breath Sound module includes Tracheal, Vesicular, Bronchial-Vesicular, Wheezes, Monophonic Wheeze, Pleural Friction Rub, Stridor, Cavernous, Crackles, Pulmonary Edema and Puppy.
Heart Sound module includes Atrial Fib, Mitral Regurgitation, Mitral Valve Click, Normal Heartbeat, PDA, Pulmoic Stenosis, Respiratory Crackles and MR Murmurs, SAS, Mitral Regurgitation, VPC and VSD.
Ordering information: This item is made to order. Returns cannot be accepted.