LaMotte® InstaTest® Water Test Strips
Supplier: LA MOTTE CHEMICAL PROD CO SE
2997 2996 3021-G-ENV
470112-656PK 26.15 CAD
470112-656 470112-662 470222-300
LaMotte® InstaTest® Water Test Strips
Environmental Test Kits
Keep it simple with these easy-to-use, individual water test strips
  • Easy to Use
  • No Additional Reagents Needed
A great way to monitor a variety of common water quality factors without having to measure reagents. Compare your readings to a handy color comparator chart printed right on the storage vial.