Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Supplier: MilliporeSigma
1.99006.0001
1.99006.0001
1.99021.0001
1.99064.0001
EM1.99005.0001EA
196.47
CAD
EM1.99005.0001
CA1.99006.0001
CAEM1990060001
EM1.99019.0001
EM1.99021.0001
EM1.99064.0001
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Buffers
pH Calibration Buffers
30 ml buffers sachets - ideal for the calibration of pH meters.
- Practical 30 mL sachets – one use per sachet – no risk of contamination
- Solutions are always fresh and ready to use
- Batch traceable to NIST
- Reference temperature: 25°C
- Precision: ±0.015 pH units max.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...