Chemizorb® Hg, Absorbent for Mercury, Merck
Supplier: Merck
1.01569.0001 1.12576.0001 1.12576.0001 1.01569.0001
CA1.12576.0001CS 8211.66 CAD
EM1.01569.0001 CA1.12576.0001 CA1.01569.0001 EM1.12576.0001
Chemizorb® Hg, Absorbent for Mercury, Merck
Spill Control Materials
Chemizorb® Hg is an all-inclusive set of reagents and auxiliaries for the safe and complete removal of drops of mercury and of traces of elemental mercury.
  • Sufficient for the safe and simple decontamination of an area of roughly one square metre
  • Chemizorb Hg does not affect skin, gloves should be worn to avoid contact with mercury
  • Mercury containing waste material must be disposed of as special waste in accordance with the official regulations
Merck products are not available from Avantor in all countries. Please check with your local distributor.
Ordering information: Pack contains 500 g reagent 1 and 100 ml reagent 2, plus accessories in plastic case.