Absorbent, Chemizorb® Granules, EMD Millipore
Supplier: EMD Millipore
1.01568.9021 1.01568.1000 1.01568.1000 1.01568.5000 1.01568.5000 1.01568.9021
CA1.01568.9021EA 3546.8 CAD
CA1.01568.9021 CA1.01568.1000 EM1.01568.1000 CA1.01568.5000 EM1.01568.9021 EM1.01568.5000
Absorbent, Chemizorb® Granules, EMD Millipore
Spill Control Materials
Chemizorb® is a coarse grained material based on chemically inert porous mineral or synthetic copolymers (grain size >1 mm (95%), bulk density approx. 400 kg/m³). It is insoluble in water and in all other media that are liquid at room temperature. Both Chemizorb® granules and powder are suited for absorbing alkalis, acids and also heating bath liquids.
  • Able to absorb up to 100% of its own weight
  • Low viscosity liquids are absorbed instantaneously, highly viscous media (e.g. glycerine, oils) within a few minutes
  • Good dosing is possible thanks to the coarse grained format
  • Granule form is used in all those places where it is not suitable to use the powder form, for example in draughty rooms or outside
Merck products are not available from Avantor in all countries. Please check with your local distributor.