Diphosphorus pentoxide, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.00570.0100 1.00570.0500
CA1.00570.0100CS 2805 CAD
CA1.00570.0100 CA1.00570.0500
Diphosphorus pentoxide, Supelco®
di-Phosphorus pentoxide
Formula: P₂O₅
Melting Pt: 562 °C
Density: 2.253 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR