Silica gel, granules desiccant, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.01907.1000 1.01907.5000
CA1.01907.1000CS 571 CAD
CA1.01907.1000 CA1.01907.5000
Silica gel, granules desiccant, Supelco®
Silica gel
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR