Silica gel, granules, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.01905.1000 1.01905.1000
CA1.01905.1000CS 2427.33 CAD
EM1.01905.1000 CA1.01905.1000
Silica gel, granules, Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR