Calcium chloride, anhydrous ~90% (by complexometric analysis), granules, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

M-Clarity™ quality level = MQ200

1.02392.1000 1.02392.5000 1.02392.9025
CA1.02392.1000CS 2023 CAD
CA1.02392.1000 CA1.02392.5000 CA1.02392.9025
Calcium chloride, anhydrous ~90% (by complexometric analysis), granules, Supelco®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.15 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR