Calcium chloride, anhydrous, granules, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
1.02379.1000
1.02379.5000
CA1.02379.1000CS
2893
CAD
CA1.02379.1000
CA1.02379.5000
Calcium chloride, anhydrous, granules, Supelco®
Calcium chloride
Formula:
CaCl₂ Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 772 °C Density: 2.174 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 REACH: 01-2119494219-28 Merck Index: 14,01659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...