VWR® Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Supplier: VWR International
10124-020 10124-022 10124-024 10124-026
10124-020CS 1046.87 CAD
10124-020 10124-022 10124-024 10124-026
VWR® Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Made from borosilicate 3.3 expansion glass.
  • Borosilicate glass construction
  • Individually serialized
  • Glass penny head stopper
  • Amber glass recommended for UV light-sensitive samples
Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.