n-Pentane ≥95% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pentane

M-Clarity™ quality level = MQ200

1.07176.5000 1.07176.5000 1.07176.9190 1.07176.1000 1.07176.9190 1.07176.1000
CA1.07176.5000CS 3893.29 CAD
CA1.07176.5000 EM1.07176.9190 CA1.07176.9190 CA1.07176.1000 EM1.07176.5000 EM1.07176.1000
n-Pentane ≥95% (by GC), EMPLURA®, Supelco®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (lit.)
Melting Pt: −130 °C (lit.)
Density: 0.626 g/ml (25°C) (lit.)
Flash Pt: –40 °C
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
Merck Index: 13,07193
https://ca.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg

Order Now

Specification Test Results

Purity (GC) ≥ 95.0 %
Identity (IR) conforms
Free acid (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/ 4 °C) 0.626 - 0.628
Readily carbonizable substances conforms
Evaporation residue ≤ 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR