VWR® Low Actinic Volumetric Flasks with PE Stoppers, Amber Glass, Class A, Serialized
Supplier: VWR
VWR®
131.276.02A 131.276.06A
10124-204CS 1371.53 CAD
10124-204 10124-202
VWR® Low Actinic Volumetric Flasks with PE Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Class A, serialized, wide-mouth, low actinic volumetric flasks are supplied with polyethylene stoppers.
  • Wide mouth
  • Amber glass fabrication
  • Polyethylene stoppers
Ordering information: Flasks are individually certified.