Laboratory Bottles, Narrow Mouth, with Thread, BrandTech
Supplier: Brandtech
704006 704008 704004
89136-834EA 94.17 CAD
89136-834 89136-836 89136-832
Laboratory Bottles, Narrow Mouth, with Thread, BrandTech
Bottles Glass Bottles
Soda-lime glass (amber).
  • Suitable for Dispensette®, seripettor® and other bottletop dispensers
  • Amber soda-lime glass
  • Pouring ring and screw cap included
The ethylene-acrylate coating envelops the glass bottle like a protective skin. Should the bottle break, the splintering hazards are reduced considerably. The maximum operating temperature for coated bottles is 80 °C.
Note: To preserve the coating, do not clean at temperatures exceeding 60 °C.
Screw cap PP, pouring ring PE-LD. Space-saving square base. The bottles are available ethylene-acrylate coated and uncoated.