Ammonium acetate ≥96.0% (by alkalimetry), EMPLURA®, Supelco®
Supplier: MilliporeSigma
Synonyms:
Acetic acid ammonium salt
M-Clarity™ quality level = MQ200
1.01115.5000
1.01115.1000
1.01115.1000
1.01115.5000
CA1.01115.5000CS
2313.1
CAD
CA1.01115.5000
CA1.01115.1000
EM1.01115.1000
EM1.01115.5000
Ammonium acetate ≥96.0% (by alkalimetry), EMPLURA®, Supelco®
Ammonium acetate
Formula:
H₃CCOONH₄ MW: 77.08 g/mol Melting Pt: 114 °C Density: 1.17 g/cm3 (20 °C) Flash Pt: 136 °C |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 UN: 3077 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...