Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
601500-0545 601500-0429 601500-0529 601500-0824 601500-0829 601500-1024 601500-0324 601500-0424 601500-0524 601500-0624 601500-0929
46200-630EA 186.3 CAD
46200-630 KT601500-0124 KT601500-0429 KT601500-0529 KT601500-0824 KT601500-0829 KT601500-0845 KT601500-1024 KT601500-2124 KT601500-0224 KT601500-0324 KT601500-0424 KT601500-0524 KT601500-0624 KT601500-0929
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Flasks Flat Bottom Flasks
Single neck flat bottom flask with a Standard Taper outer joint.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.