VWR® Micro Volumetric Flasks with Glass Stoppers, Class A, Serialized
Supplier: VWR International
10124-362 10124-370 10124-378 10124-388
10124-362CS 2583.77 CAD
10124-362 10124-370 10124-378 10124-388
VWR® Micro Volumetric Flasks with Glass Stoppers, Class A, Serialized
Flasks Volumetric Flasks
Clear glass volumetric flasks are calibrated to contain at 20°C and comply to ASTM E237 standards.
  • Clear siding
Flasks are seralized and supplied with one glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.