Rod Mounted Swing Latch Clamps, Ace Glass
Supplier: Ace Glass
6442-02 6442-04 6442-06
10098-916EA 1402.48 CAD
10098-916 10098-918 10098-920
Rod Mounted Swing Latch Clamps, Ace Glass
Clamps General Purpose Clamps
Swing latch clamp secures neck of Protean Fusion reaction vessel to the stand.
May also be used to support any round object with an equivalent maximum outside diameter. Stainless steel rod support is 1/2" O.D×8" long.