Accessories for Whatman™ Vacuum Filtration Apparatus, Witt's Flask, Whatman products (Cytiva)
Supplier: Cytiva
10477600 10471700 10464103 10477601 10464100 10464103 10471700 10477600 10477601
CA11013-202PK 217.25 CAD
CA11013-202 CA11013-512 CA11013-514 CA11013-516 10035-200 10035-202 10035-206 10035-208 10035-210
Accessories for Whatman™ Vacuum Filtration Apparatus, Witt's Flask, Whatman products (Cytiva)
Manifolds
Witt's bottle for filtrate collection in an inserted container. Borosilicate glass bottle, 100 x 160 mm, with replaceable round lid and side-mounted tubing nozzle for vacuum tubing 8 mm (inside diameter).
  • AC model
  • Contamination-free pumping of air, gases and vapours
  • Extremely quiet and smooth running
  • Equipped with thermo switch and standard fuse
  • Oil-free membrane pump
  • Delivery m³/h: 3.6 l/min, vacuum: <100 mbar absolute, pressure: 4 bar, weight: 11 kg