Hydrion® Double Roll pH Paper, Micro Essential Laboratory
Supplier: Micro Essential
161 150 151 3-8 193
13796-106CS 281.95 CAD
13796-106 60776-002 60777-049 60778-041 73450-010
Hydrion® Double Roll pH Paper, Micro Essential Laboratory
Test Papers pH Indicator Papers and Test Strips
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability.
Double Roll pH Papers consist of two rolls of pH papers packaged in a protective plastic dispenser with stainless steel cutoff lips and two matching color charts
Double Roll pH Wide Range Papers have a clear color match at each full pH unit: [0.0][1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0][12.0][13.0][14.0]
pH color changes are distinct and easily matched to the color chart.
Ordering information: Each carton (case) contains 10 dispensers, each consisting of two (2) 15-foot rollsMore Product Information