Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
M-Clarity™ quality level = MQ100
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 Melting Pt: -45.7 Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Purity (GC) | ≥ 99.9 % |
Identity (IR) | conforms |
Evaporation residue | ≤ 1.0 mg/l |
Water | ≤ 0.01 % |
Color | ≤ 10 Hazen |
Acidity | ≤ 0.0001 meq/g |
Alkalinity | ≤ 0.0002 meq/g |
Al (Aluminium) | ≤ 10 ppb |
Ca (Calcium) | ≤ 10 ppb |
Fe (Iron) | ≤ 10 ppb |
K (Potassium) | ≤ 5 ppb |
Mg (Magnesium) | ≤ 10 ppb |
Na (Sodium) | ≤ 50 ppb |
Any further metal (ICP-MS) | ≤ 5 ppb |
Gradient grade (at 210 nm) | ≤ 1.0 mAU |
Gradient grade (at 254 nm) | ≤ 0.5 mAU |
Fluorescence (as quinine at 254 nm) | ≤ 1.0 ppb |
Fluorescence (as quinine at 365 nm) | ≤ 0.5 ppb |
Transmission (at 191 nm) | ≥ 25 % |
Transmission (at 195 nm) | ≥ 85 % |
Transmission (at 200 nm) | ≥ 96 % |
Transmission (at 215 nm) | ≥ 98 % |
Transmission (from 230 nm) | ≥ 99 % |
Suitable for LC-MS; Intensity of background mass peak based on reserpine (APCI/ESI positive) | ≤ 2 ppb |
Suitable for LC-MS; Intensity of background mass peak based on reserpine (APCI/ESI negative) | ≤ 20 ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...