KeepIT® Freezer Boxes, Low-Profile, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W651702-R W651702-P W651702-Y W651702-W W651702-G W651702-B W651703-R W651703-P W651703-W W651703-Y W651703-G W651703-B W651704-R W651704-P W651704-W W651704-Y W651704-G W651704-B
89495-472CS 237.46 CAD
89495-472 89495-474 89495-476 89495-478 89495-480 89495-482 89495-484 89495-486 89495-488 89495-490 89495-492 89495-494 89495-496 89495-498 89495-500 89495-502 89495-504 89495-506
KeepIT® Freezer Boxes, Low-Profile, Wheaton
Boxes Cryoboxes
The WHEATON KeepIT® Freezer Boxes provide an ideal method for batching and storing samples
The KeepIT-25 accomodates up to 25 external threaded vials. The KeepIT-81 accomodates up to 81 external threaded and the KeepIT-100 accomodates up to 100 internal threaded vials (1.2 – 2 mL).
Openings for each vial in the bottom of the box facilitate the scanning of WHEATON CryoELITE 2D Data Matrix Bar Code Inserts on CryoELITE vials. Made from Eastman Tritan™ BPA free and shatter resistant resin. Boxes have a standard footrpint that is compatible with liquid nitrogen storage shelves and freezerMore Product Information