PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
Supplier: Corning
5635-25 5635-50 5635-200 5635-10 5635-100 5635-250
22877-016CS 492.88 CAD
22877-016 22877-018 33500-514 33500-516 33500-518 33500-520 66025-672 66025-682 77777-081 77777-083
PYREX® EZ Access™ Volumetric Flasks, Class A, Corning
Flasks Volumetric Flasks
PYREX® glass, low actinic, with stopper.
  • Graduation line is sharp and permanent
  • Low actinic glass provides protection for materials sensitive to light
  • Chemical and thermal resistance
Ideal for microchemical work. Features wide mouth facilitates pipete filling and mixing.
The flasks are supplied with larger interchangeable glass stoppers
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey