Porcelin Combustion Boats, CoorsTek
Supplier: CoorsTek
60032
22825-104CS 2143.48 CAD
22825-104
Porcelin Combustion Boats, CoorsTek
Boats Combustion Boats
Combustion boats are glazed inside and out, except for the exterior bottom surface
Boats feature a perforated handle, and will withstand temperatures up to 1150°C. To prevent thermal stress cracks on porcelain ware, CoorsTek strongly recommends a heating/cooling rate not to exceed 200°C/hour.