Whatman™ Regenerated Cellulose Membranes, Circles, Whatman products (Cytiva)
Supplier: Cytiva
10410012 10410206 10410212 10410219 10410224 10410312 10410319 10410229 10410214 10410380 10410014 10410314 10410014 10410214 10410229 10410314 10410380 10410012 10410206 10410212 10410219 10410224 10410312 10410319
97010-094PK 511.84 CAD
97010-094 97010-096 97010-098 97010-100 97010-102 97010-104 97010-106 CA11012-912 CA11013-042 CA11013-246 CA11013-502 CA11013-520 10034-990 10034-992 10034-994 10034-996 10034-998 CA97010-094 CA97010-096 CA97010-098 CA97010-100 CA97010-102 CA97010-104 CA97010-106
Whatman™ Regenerated Cellulose Membranes, Circles, Whatman products (Cytiva)
Filters Membrane Filters
Cytiva's Whatman regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
  • Spontaneous wetting with very good wet strength
  • Chemical resistance suitable for a wide range of aqueous and organic media
  • Mechanically stable with low protein binding
  • Low extractable levels to minimize sample contamination
  • Can be sterilized by all common methods