KIMBLE® CHROMAFLEX® Column End Fittings with Bed Support, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
420832-4110 420832-2100 420832-2110 420832-2120 420832-4120
KT420832-4110EA 201.36 CAD
KT420832-4110 KT420832-2100 KT420832-2110 KT420832-2120 KT420832-4120
KIMBLE® CHROMAFLEX® Column End Fittings with Bed Support, DWK Life Sciences
Fittings and Connectors
End fittings with bed support to go on the bottom of the CHROMAFLEX® columns
The 2000 series is PTFE and is to be used with tapered and standard columns, the 4000 series to be used with jacketed columns and the 5000 series to be used with flow adapters.
Ordering information: Fittings require CTFE adapter, available separately.