VWR® Model 1000 & 2000 Advanced Large Capacity Stirrers, 230 V (Export Only)
Supplier: VWR International
12621-060 12621-064
12621-060EA 8092.11 CAD
12621-060 12621-064
VWR® Model 1000 & 2000 Advanced Large Capacity Stirrers, 230 V (Export Only)
Stirrers Magnetic Stirrers
Ideal for high volume applications, the powerful magnetic drive of this stirrer is capable of mixing high viscosity materials.
  • 230 V model
  • LED displays for speed and time
  • Durable stainless steel construction
  • Best-in-Class Five-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Stainless steel base offers durability and added stability.
Microprocessor control precisely regulates speed through the entire range. Stirring function, with continuous duty motor and magnet, maintains set speed even under changing load or viscosity. Speed ramping slowly increases speed for improved safety and enhanced coupling. Avoids splashing, improves spin bar control, and provides excellent low end speed control.
Certifications: RoHS Certified. TUV listed. CE Marked. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 234 cm (92") Euro type plug. Unit is also supplied with a 7 cm (2.75") egg-shaped PTFE-coated stir bar. Stirrers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
Caution: 230 V model.