Scale Up Series™ Jacketed Cylindrical Reactor Flasks, Ace Glass
Supplier: Ace Glass
6441-02 6441-04 6441-06 6441-08 6441-10 6441-12 6441-14 6441-16 6441-18
10098-898EA 2132.26 CAD
10098-898 10098-900 10098-902 10098-904 10098-906 10098-908 10098-910 10098-912 10098-914
Scale Up Series™ Jacketed Cylindrical Reactor Flasks, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
Scale-Up Series™ Jacketed Cylindrical Flasks are equipped with an Easy Drain glass valve and KF/NW inlet and outlet
The flask is graduated on both sides, which allows for either left- or right-handed flask orientation. The glass plug Easy Drain valve is designed to prevent accidental valve removal when opening, and incorporates a compression tube angled side arm for maximum unobstructed drainage. Flasks are available in sizes ranging from 100mL to 6000mL. Operating temperature limits are –60 to 200°C (–76 to 392°F).