Graduated Reusable Centrifuge Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
410551-0005 410551-0013
KT410551-0005EA 154.04 CAD
KT410551-0005 KT410551-0013
Graduated Reusable Centrifuge Tubes, Kimble Chase
Tubes Centrifuge Tubes
Replacement tubes for the 410550 series graduated reusable centrifuge tubes with flat head stoppers.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.