Medium Length Hex Head Hollow Glass Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
850400-1420 850400-1922 850400-2425 850400-2926
KT850400-1420EA 70.53 CAD
KT850400-1420 KT850400-1922 KT850400-2425 KT850400-2926
Medium Length Hex Head Hollow Glass Stoppers, Kimble Chase
Stoppers Glass Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders, and separatory funnels
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Hollow, hexagonal head medium length Standard Taper stopper can be easily gripped, and may be set on the flattened top to avoid contamination of the ground surface.