Whatman™ Standard Cellulose Extraction Thimbles
Supplier: Cytiva
10350240 10350220 10350242 10350217 10350220 10350242 10350217 10350274 10350287 10350274 10350287 10350261 10350324 10350223 10350225 10350238 10350219 10350241 10350273 10350255 10350250 10350437 10350234 10350252 10350227 10350247 10350211 10350226 10350265 10350243 10350215 10350306 10350275 10350236 10350245 10350219 10350223 10350225 10350226 10350227 10350234 10350236 10350238 10350241 10350243 10350245 10350247 10350250 10350252 10350255 10350261 10350265 10350273 10350275 10350306 10350324 10350437 10350211 10350215 10350240
28152-844PK 172.77 CAD
28152-844 CA28152-846 CA28152-848 CA74330-280 28152-846 28152-848 74330-280 CA89233-656 CA89233-658 89233-656 89233-658 CA11013-416 CA11013-426 CA11013-440 CA11013-444 CA11013-448 CA11013-450 CA11013-458 CA11013-460 CA11013-462 CA11013-528 CA11013-530 CA11013-532 CA11013-536 CA11013-538 CA11013-540 CA11013-546 CA11013-550 CA11013-556 CA11013-558 CA11013-560 CA11013-834 CA11013-842 CA11013-844 CA11013-846 10034-778 10034-780 10034-782 10034-784 10034-786 10034-788 10034-790 10034-792 10034-794 10034-796 10034-798 10034-800 10034-802 10034-804 10034-806 10034-808 10034-810 10034-812 10034-814 10034-816 10034-818 10034-820 10035-876 10035-878 CA28152-844
Whatman™ Standard Cellulose Extraction Thimbles
Extractors Extraction Thimbles
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.