Ammonium persulfate (APS), powder

Supplier: G-Biosciences

Synonyms: Ammonium persulfatePeroxidisulphuric acid diammonium saltAmmonium persulphatePeroxidisulfuric acid diammonium salt

P011-B P011-B
CA71003-316EA 84.4 CAD
71003-316 CA71003-316
Ammonium persulfate (APS), powder
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00003390
CAS Number: 7727-54-0
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://ca.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR