D-(+)-Biotin

Supplier: Biotium

Synonyms: Vitamin HVitamin B85-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acidD-BiotinVitamin B7

90072
89139-648EA 91.37 CAD
89139-648
D-(+)-Biotin
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00005541
CAS Number: 58-85-5
Merck Index: 13,01231

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR