IPTG (isopropyl-β-D-thiogalactopyranoside)

Supplier: Biotium

10021 10021-1 10021-2
71004-000EA 72.46 CAD
71004-000 71004-002 71004-004
IPTG (isopropyl-β-D-thiogalactopyranoside)
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR