4-Methylumbelliferyl phosphate

Supplier: Biotium

Synonyms: Phosphoric Acid 4-Methylumbelliferyl Ester, 4-Methylumbelliferyl phosphoric acid, 4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphate

10009 10009-1
89138-208EA 134.16 CAD
89138-208 89138-210
4-Methylumbelliferyl phosphate
4-Methylumbelliferyl phosphate
Formula: C₁₀H₉O₆P
MW: 256.15 g/mol
MDL Number: MFCD00016969
CAS Number: 3368-04-5
EINECS: 222-137-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR