Nitro blue tetrazolium chloride (NBT)

Supplier: Biotium

Warning

Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

10008 10008-1 10008-2
89138-202EA 86.47 CAD
89138-202 89138-204 89138-206
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR