5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)

Supplier: Biotium

Warning

Synonyms: BCIP, P-toluidine 5-bromo-4-chloro-1H-indol-3-yl phosphate

10002 10002-2
89138-186EA 130.54 CAD
89138-186 89138-190
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formula: C₁₅H₁₅BrClN₂O₄P
MW: 433.6214 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00040636
CAS Number: 6578-06-9
EINECS: 229-506-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR