5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Supplier: Biotium

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

10001 10001-2
89138-180EA 133.81 CAD
89138-180 89138-184
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR