Eisco® NextGen Bunsen Burner with Flame Stabilizer and Gas Adjustment
Supplier: Wards
CH0993NG CH0993LP
470121-700EA 61.15 CAD
470121-700 470121-702
Eisco® NextGen Bunsen Burner with Flame Stabilizer and Gas Adjustment
Heaters Gas Burners
The next generation in laboratory burners. NextGen Bunsen Burners are designed to be the safest on the market.
  • Designed for safety
  • Expanded cast iron base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Candle stick style handle allows user to move burner without getting harmed
  • Features air flow adjustment sleeve and brass flame stabilizer
  • Base is coated with a chemical resistant powder coating
This versatile Bunsen burner produces a safe continuous stream of flammable gas used for heating, sterilization and combustion. The burner features an air flow adjustment sleeve, brass gas adjustment knob, and brass flame stabilizer. This is a very safe and easy to use burner. Ribbed fuel input tube is 8 mm outside diameter, and versions jetted for liquid propane and natural gas are both available. Liquid propane burners have a BTU range of approx. 2000 to 3200. Maximum Temperature is approximately 1550 °C (2822° F). Natural gas burners have a BTU range of approx. 800 to 1200. Maximum temperature is approximately 1500 °C (2732° F).
Ordering information: Neoprene connecting hoses, 60 or 90 cm (items 470121-742 and 470220-806), recommended.