CryoELITE® Cryogenic Vials with Pre-Inserted Barcodes, Freestanding, External Thread, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W985864-BC W985865-BC W985866-BC W985867-BC W985868-BC W985863-BC
10147-448CS 1054.68 CAD
10147-448 10147-450 10147-452 10147-454 10147-460 10147-772
CryoELITE® Cryogenic Vials with Pre-Inserted Barcodes, Freestanding, External Thread, Wheaton
Vials
Wheaton CryoELITE® Cryogenic Freestanding Vials have a Loctagon™ Vial Skirt that provides stability in the freestanding position and locks into the CryoELITE® Benchmate Rack in order to provide easy open and close with one hand
The 2D Data Matrix Bar Code Bottom Inserts allows for immediate bar coding of samples. The 2D Data Matrix Bar Code Insert is locked into place in the bottom of the vial. The insert is made of the same polypropylene as the vials themselves and is laser-etched for durability. 
These vials are supplied with Wheaton 2D Data Matrix Bar Codes inserted. The BarMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable