Mouse Recombinant FGF Basic
Supplier: IBI Scientific
RPM-200 RPM-201 RPM-202
10039-908EA 325.83 CAD
10039-908 10039-910 10039-912
Mouse Recombinant FGF Basic
Proteins and Peptides
IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.
IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.
The recombinant mouse protein FGF Basic has a gene ID of 14173. The calculated molecular weight is 16.2kDa.