Cryoscope Standards, Advanced Instruments
Supplier: ADVANCED INSTRUMENTS INC.
3LA022 3LA023 3LA033
52198-144EA 111.3 CAD
52198-144 55005-001 55005-003
Cryoscope Standards, Advanced Instruments
Standards Environmental Analysis Standards
Cryoscope standards are used to calibrate and/or check the performance of a cryoscope.
Standards should only be used in accordance with the cryoscope being calibrated. Refer to the specific instrument's user guide for calibration instructions. These standards are prepared from salts. 2-Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01%, is added as a preservative.