Adhesive Crystallography Seal, Thermo Scientific
Supplier: Thermo Scientific
AB-2450
89499-576CS 14572.06 CAD
89499-576
Adhesive Crystallography Seal, Thermo Scientific
Seals Microplate Seals
Optically clear, pressure sensitive, non-sticky adhesive seal for crystallography and genotyping applications.
Samples are not exposed to adhesive. Seal is easily peelable without leaving residue on the plate. Rolls are designed for use with the Thermo Scientific SEAL-IT 100™ adhesive sealer.