Hanks' Balanced Salt Solution (HBSS), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
114-052-101 114-061-101 114-062-101 119-065-101
10128-666EA 66.71 CAD
10128-666 10128-740 10128-742 10128-838
Hanks' Balanced Salt Solution (HBSS), Quality Biological
Buffers Cell Culture Buffers
Hanks' Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. HBSS provides cells with a source of water and essential inorganic ions, and carbohydrate as an energy source.
HBSS without Ca and Mg can also be useful for diluting cells for counting
Formulations with calcium and magnesium support cell adhesion and are generally used as transport media, or for reagent preparation. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
While HBSS with Ca & Mg is useful for transporting cells or tissue samples, preparing reagents, suspension cultures, as well as to resolve clumping when clumping is a problem.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.