Leibovitz' L-15 Medium, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
112-029-101
10128-632EA 121.31 CAD
10128-632
Leibovitz' L-15 Medium, Quality Biological
Cell Culture Media
Most media are buffered by a combination of sodium bicarbonate and an atmosphere enriched in CO2. The latter is usually provided through the use of a specially equipped incubator. L-15 was developed at a time when such equipment was not readily available. Dr. Leibovitz developed a buffering system based on free base amino acids, inorganic salts and the substitution of galactose for glucose.
  • Without Sodium Bicarbonate
  • Designed for cell culture without CO2
  • With Phenol Red
  • Without L-glutamine
L-15 has been used for the culture of HEp-2 monkey kidney and primary explants of embryonic and adult human tissue, and a variety of other cells. Many viruses have also been cultivated in this medium.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only