KIMBLE® TFE/Propylene O-Rings, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
758260-0027 758260-0118
KT758260-0027CS 78.43 CAD
KT758260-0027 KT758260-0118
KIMBLE® TFE/Propylene O-Rings, DWK Life Sciences
Seals Sealing Rings
These O-rings are used for replacement parts in stopcocks, valves, chromatography columns, and closures.
Not recommended for use with aromatics, non-polar solvents, ethers, ketones, freon, acetic acid and some chlorinated solvents. Temperature service range is -45 °C to 204 °C with some short term use to 316 °C and density: 1.68.
O-ring copolymer is composed of tetrafluoroethylene/propylene and has excellent physical and mechanical properties. Resistant to alcohols, acids, bases, oxidizing agents, oils, lubricants and many solvents.