Flanged Plug Caps, Simport Scientific
Supplier: Simport Scientific
T404-3B T404-3R T404-3W T404-3Y T404-10B T404-10G T404-10N T404-10W T404-10R T404-10Y T404-3N T404-3G
CA60831-001PK 77.25 CAD
CA60831-001 CA60831-002 CA60831-004 CA60831-006 CA60831-007 CA60831-008 CA60831-009 CA60831-011 CA60831-012 CA60831-013 CA60831-003 CA60831-010
Flanged Plug Caps, Simport Scientific
Closures Test/Sample Tube Closures
This line of polyethylene flanged plug caps provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.
Flanged plug caps feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. Available in a variety of colors to facilitate identification.