KIMBLE® Inner Joint Hose Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
182900-2440 276100-1922
KT182900-2440EA 112.07 CAD
KT182900-2440 KT276100-1420 KT276100-1922
KIMBLE® Inner Joint Hose Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
This inlet adapter is ideal for use as a vent, or for the introduction of pressure/vacuum.
  • Inner [ST] joint at one end
  • Vertical hose connection at other end
  • Manufactured from borosilicate glass
Adapter features an inner Standard Taper joint at one end and vertical hose connection at the other.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable