Educational-Grade Burets, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
17021H-25 17021H-50
89001-598CS 1597.5 CAD
89001-598 89001-600 75799-984
Educational-Grade Burets, Kimble Chase
Burettes
These economical and versatile burets are ideal for use in educational institutions and for many general laboratory procedures.
  • Funnel fill style
  • Easy-to-read durable black enamel scale
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The 17021H burets are supplied with a PTFE compression fit stopcock assembly and a precision ground tip. The 17040K buret is supplied with a PTFE stopcock plug.
Ordering information: The KIM-KAP™ dust cap is included.