Industrial-Grade Boot Covers, Yuleys Clean Step System, HexArmor
Supplier: HexArmor
32001-A 32001-B 32001-C 32001-D 32001-E 32001-F 32001-G 32001-H 32001-I
10147-402PR 209.13 CAD
10147-402 10147-404 10147-406 10147-416 10147-418 10147-420 10147-422 10147-424 10147-426
Industrial-Grade Boot Covers, Yuleys Clean Step System, HexArmor
Workwear
The hands-free design of these HexArmor boot covers allows you to easily slip them on and off without bending over or using your hands.
  • Hands-free on-and-off usage
  • Slip-reducing tread design
  • Heel-locking grip system
  • Autoclavable
  • Superior chemical and abrasion resistance
  • Greater flexibility at extreme temperatures
  • Silicone free
The Yuleys Clean Step System boot covers are chemical and abrasion-resistant
The Yuleys Clean Step System boot covers have a high temperature resistance. They are anti-static, autoclavable, bleachable and launderable. The Yuleys Clean Step System boot covers offer excellent grip on dry surfaces.
They are also moderately oil-resistant.